Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-Barbituric Acid Reagent, For Cyanide, Certified, LabChem™

Catalog No. LC221901 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC221901 500 mL
LC221902 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC221901 Supplier LabChem, Inc. Supplier No. LC221901
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Pyridine-Barbituric Acid Reagent
Color Amber
Name Note For Cyanide
CAS Max % 62, 30, 5.6, 2.4
CAS 110-86-1, 67-52-7, 7647-01-0
Linear Formula C5H5N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 79.1
Grade Certified
Identification Passes Test
Packaging Amber Glass
Suitability Passes Test
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Hydrogen chloride
Quantity 500 mL
Physical Form Liquid
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.