Learn More
Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide), Reagents
Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide), Reagent, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F). Manufactured in ISO 9001 facility.
Supplier:  Reagents Holdings Llc CP302600500C
 View more versions of this product
Catalog No. 01-334-095
 Restricted Item
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide)
7732-18-5 , 110-86-1 , 67-52-7
Reagent
1.0654
Non-sterile
Yellow
100°C
Glass Bottle
500 mL
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories