Learn More
Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide), Reagents
Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide), Reagent, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F). Manufactured in ISO 9001 facility.
$64.20 - $216.00
Specifications
| Chemical Name or Material | Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide) |
|---|---|
| Color | Yellow |
| CAS | 7732-18-5 , 110-86-1 , 67-52-7 |
| Boiling Point | 100°C |
| Grade | Reagent |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-334-097
|
Reagents Holdings Llc
CP302600120C |
120 mL |
Each for $64.20
|
|
|||||
|
01-334-096
|
Reagents Holdings Llc
CP3026001C |
1 L |
Each for $216.00
|
|
|||||
|
01-334-095
|
Reagents Holdings Llc
CP302600500C |
500 mL |
Each for $162.00
|
|
|||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.