Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide), Reagents

Catalog No. 01334097
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-097 120 mL
01-334-096 1 L
01-334-095 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-097 Supplier Reagents Holdings Llc Supplier No. CP302600120C
Add to Cart
Add to Cart

Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide), Reagent, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Pyridine-Barbituric Acid Stock Solution (APHA for Cyanide)
Color Yellow
CAS 7732-18-5, 110-86-1, 67-52-7
Boiling Point 100°C
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0654
Quantity 120 mL
Sterility Non-sterile
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.