Learn More
Pyridine-d{5}, 99.5% (Isotopic), contains 0.05% v/v TMS
Description
Pyridine-d{5} is used in dehalogenation reactions and can be used as a base in condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 5 g |
| Density | 1.05 g/mL |
| Odor | Unpleasant |
| Flash Point | 20°C (68°F) |
| Refractive Index | 1.506 |
| Melting Point | -42°C |
| Boiling Point | 115°C |
| UN Number | UN1282 |
| Beilstein | 114377 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H225-H302+H312+H332-H315-H319 |
| missing translation for 'healthHazard2' | GHS H Statement H225-H302-H312-H332 Highly flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| missing translation for 'healthHazard3' | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE |
| EINECSNumber | 203-720-2 |
| TSCA | No |
| Recommended Storage | Ambient |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.