Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-d{5}, 99.5% (Isotopic)

Catalog No. AA3647104
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3647104 2 g
AA3647109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3647104 Supplier Thermo Scientific Chemicals Supplier No. 03647104
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7291-22-7 | C5H5N | 84.13 g/mol

Pyridine-d{5} is used in the synthesis complexes with cobalt(II) halides, pyridine-d5, a deuterated form of pyridine, ionic liquid matrices (ILMs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7291-22-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 84.13
MDL Number MFCD00044639
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
PubChem CID 558519
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

Specifications

Physical Form Liquid
Quantity 2 g
Density 1.05 g/mL
Flash Point 20°C (68°F)
Odor Unpleasant
Refractive Index 1.506
Melting Point -42°C
Boiling Point 115°C
UN Number UN1282
Beilstein 114377
Solubility Information Soluble in water,alcohol,ether,acetone,and benzene.
Formula Weight 84.14
Percent Purity 99.5%
Chemical Name or Material Pyridine-d5, Isotopic
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE
EINECSNumber 203-720-2
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.