Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyridine d5 99.5 atom D25G
Catalog No. 502903809
Encompass
Pyridine-d5 is a deuterated NMR solvent useful in NMR-based research and analyses. It has been synthesized by palladium catalyzed H/D (hydrogen/deuterium) exchange reaction between pyridine vapor and heavy water. Infrared and Raman spectra of pyridine-d5 have been recorded in the range 300-4000cm-1.
Catalog No. 50-290-3809 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 53297525G
Pyridine-d5 is a deuterated NMR solvent useful in NMR-based research and analyses. It has been synthesized by palladium catalyzed H/D (hydrogen/deuterium) exchange reaction between pyridine vapor and heavy water. Infrared and Raman spectra of pyridine-d5 have been recorded in the range 300-4000cm-1.