Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-d5, for NMR, 99.5 atom % D

Catalog No. AC464190100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464190100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC464190100 Supplier Thermo Scientific Chemicals Supplier No. 464190100
Add to Cart
This item is not returnable. View return policy

CAS: 7291-22-7 | C5H5N | 84.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7291-22-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 84.13
MDL Number MFCD00044639
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
IUPAC Name (²H₅)pyridine
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

Specifications

Flash Point 20°C
Quantity 10 mL
Refractive Index 1.5060
Specific Gravity 1.05
Density 1.0500 g/mL
Melting Point -41.0°C
Boiling Point 114.4°C
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 84.14
Grade NMR
Chemical Name or Material Pyridine-d5
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful in contact with skin.

Harmful if swallowed.

Harmful if inhaled.

Highly flammable liquid and vapor.

Causes skin irritation.

Causes serious eye irritation.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 230-720-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.