Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-d5, for NMR, 99 atom % D

Catalog No. AC166570250
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166570250 25 mL Glass Bottle
AC166570050 5 mL Glass Bottle
AC166570100 10 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC166570250 Supplier Thermo Scientific Chemicals Supplier No. 166570250
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7291-22-7 | C5H5N | 84.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7291-22-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 84.13
MDL Number MFCD00044639
InChI Key JUJWROOIHBZHMG-RALIUCGRSA-N
Synonym pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ∼2∼h_5_ pyridine, de85c
PubChem CID 558519
IUPAC Name 2,3,4,5,6-pentadeuteriopyridine
SMILES [2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]

Specifications

Physical Form Liquid
Refractive Index 1.506
Color Colorless
Flash Point 20°C
Packaging Glass Bottle
Quantity 25 mL
Deuterium (D) 98.5% min.
Water 0.1% max. (Coulometric)
Density 1.0500g/mL
Melting Point -41.0°C
Boiling Point 114.4°C
Beilstein 20, III, 2305
Formula Weight 84.14
Grade NMR
Chemical Name or Material Pyridine-d5, 99 atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-720-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.