Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, Solstice

Catalog No. 60090480 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
200 L
Packaging:
Returnable Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-480 200 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-480 Supplier Solstice Supplier No. BC333200
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

BioSyn™, for biosynthesis and applications requiring low water content, >99.8%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
PubChem CID 1049
ChEBI CHEBI:16227

Specifications

Quantity 200 L
Flash Point 20 °C
Packaging Returnable Steel Drum
Physical Form Liquid, Clear
Grade Low Water
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.