Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, Low Water, HPLC, J.T. Baker™

Catalog No. 02003865 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-865 4 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-865 Supplier Avantor Supplier No. 939303
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

High purity solvents for use in analytical and preparative separations

  • For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry
  • NOTE: Intended for nonfuel end purposes only

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Quantity 4 L
Flash Point 20°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.98kg
Formula Weight 79.1
Grade HPLC
Chemical Name or Material Pyridine, Low Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.