Learn More
Pyridine, puriss., Reag. Ph. Eur., ≥99.5% (GC), Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Solstice has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| pH | ∼8.8 (at 20°C) |
| Melting Point | -42°C |
| Color | Colorless |
| Density | 0.978g/mL at 25°C |
| Quantity | 6 x 1 L |
| Explosion Limit | 0.124 |
| Flash Point | 17°C (62.6°F) |
| Ignition Point | 899°F |
| Odor | Unpleasant |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P305 + P351 + P338 |
| DOTInformation | class 3 - PG 2 - Pyridine |
| RTECSNumber | 203-809-9 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.