Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605981 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-981 4 x 4 L Amber Glass Bottle
18-605-973 100 mL Amber Glass Bottle
18-605-975 500 mL Amber Glass Bottle
18-605-977 1 L Amber Glass Bottle
18-605-976 6 x 500 mL Amber Glass Bottle
18-605-980 4 L Amber Glass Bottle
18-605-978 6 x 1 L Amber Glass Bottle
18-605-974 20 L Steel Can
18-605-979 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-605-981 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P14504LTGLCS4
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1450, 110-86-1, C5H5N

Spectrum™ Chemical Pyridine, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.10
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

CAS Max % 1
Quantity 4 x 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 2e-005
Water 0.001
Boiling Point 115°C
Percent Purity 99%
Grade Reagent
Chemical Name or Material Pyridine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.