Learn More
Medchemexpress LLC Pyridinium, 3-(aminocarbonyl)-1-β-D-ribofuranosyl-, chloride (1:1) | 23111-00-4 | MFCD27952781 | 99.6% | 290.70 g/mol | C11H15ClN2O5 | 100G
SDP
Catalog No. 5000235531
Encompass_Preferred
Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, a vitamin B3-derived NAD+ precursor provided as a research-grade white to off-white powder for biochemical and metabolic studies. It is used to elevate cellular NAD+ levels and to investigate sirtuin activation, energy metabolism, and aging-related pathways.
  • Increases cellular NAD+ levels.
  • Activates sirtuin proteins SIRT1 and SIRT3.
  • Chloride salt form with good aqueous solubility.
  • Suitable for in vitro and in vivo metabolic research.
  • High-purity, batch-verified material for reproducible results.
Catalog No. 50-002-35531 Supplier Medchemexpress LLC Supplier No. HY123033A100G
Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, a vitamin B3-derived NAD+ precursor provided as a research-grade white to off-white powder for biochemical and metabolic studies. It is used to elevate cellular NAD+ levels and to investigate sirtuin activation, energy metabolism, and aging-related pathways.
  • Increases cellular NAD+ levels.
  • Activates sirtuin proteins SIRT1 and SIRT3.
  • Chloride salt form with good aqueous solubility.
  • Suitable for in vitro and in vivo metabolic research.
  • High-purity, batch-verified material for reproducible results.