Learn More
Medchemexpress LLC Pyridinium bisretinoid A2E TFA | 1821308-73-9 | 90.0% | 705.93 g/mol | C44H58F3NO3 | 5 MG
SDP
Catalog No. 5000222046
Encompass_Preferred
Pyridinium bisretinoid A2E TFA is the trifluoroacetate salt of A2E, a lipofuscin-derived pyridinium bisretinoid used as a research reagent in retinal studies. A2E is a fluorophore isolated from retinal pigment epithelium lipofuscin and is commonly used to model blue-light-induced phototoxicity, autophagy, and apoptosis in vitro. Supplied as a solid with reported purity and recommended storage conditions for stability.
Catalog No. 50-002-22046 Supplier Medchemexpress LLC Supplier No. HY134928A5MG
Pyridinium bisretinoid A2E TFA is the trifluoroacetate salt of A2E, a lipofuscin-derived pyridinium bisretinoid used as a research reagent in retinal studies. A2E is a fluorophore isolated from retinal pigment epithelium lipofuscin and is commonly used to model blue-light-induced phototoxicity, autophagy, and apoptosis in vitro. Supplied as a solid with reported purity and recommended storage conditions for stability.