Learn More
Medchemexpress LLC Pyridinium bisretinoid A2E (TFA) | 1821308-73-9 | 90.0% | C44H58F3NO3 | 10MG
SDP
Catalog No. 5000260157
Encompass_Preferred
Pyridinium bisretinoid A2E TFA is the trifluoroacetate salt of the lipofuscin-derived fluorophore A2E, provided as a solid research reagent for studies of retinal pigment epithelium phototoxicity, blue-light-induced apoptosis, and autophagy. It is supplied in small milligram quantities with documented purity and storage recommendations.
  • Functions as a lipofuscin-derived fluorophore for retinal research.
  • Initiates blue-light-induced apoptosis in retinal pigment epithelium models.
  • Offered with documented purity suitable for research applications.
  • Available in multiple small-scale quantities for laboratory use.
  • Stable when stored sealed at 4°C and protected from moisture and light.
  • Solvent solutions can be stored at -80°C for up to six months.
Catalog No. 50-002-60157 Supplier Medchemexpress LLC Supplier No. HY134928A10MG
Pyridinium bisretinoid A2E TFA is the trifluoroacetate salt of the lipofuscin-derived fluorophore A2E, provided as a solid research reagent for studies of retinal pigment epithelium phototoxicity, blue-light-induced apoptosis, and autophagy. It is supplied in small milligram quantities with documented purity and storage recommendations.
  • Functions as a lipofuscin-derived fluorophore for retinal research.
  • Initiates blue-light-induced apoptosis in retinal pigment epithelium models.
  • Offered with documented purity suitable for research applications.
  • Available in multiple small-scale quantities for laboratory use.
  • Stable when stored sealed at 4°C and protected from moisture and light.
  • Solvent solutions can be stored at -80°C for up to six months.