Learn More
Medchemexpress LLC Pyridinium bisretinoid A2E trifluoroacetate | 00-00-0 | 90.0% | 709.96 | C44H54D4F3NO3 | 5 MG
SDP
Catalog No. 5000222478
Encompass_Preferred
Pyridinium bisretinoid A2E-d4 TFA is a deuterium-labeled analog of the retinal bisretinoid A2E supplied as the trifluoroacetate salt for use as an isotopically labeled standard in biochemical and phototoxicity research. It models the photoactivated behavior of native A2E and is intended for quantitative analysis, tracer experiments, and studies of apoptosis and autophagy in retinal pigment epithelium.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • Supplied as a trifluoroacetate salt for handling and solubility in common organic solvents.
  • Suitable for phototoxicity, apoptosis, and autophagy research applications.
  • Certificate of analysis available reporting purity by NMR (90.0%).
  • Offered in small research pack sizes appropriate for analytical workflows.
Catalog No. 50-002-22478 Supplier Medchemexpress LLC Supplier No. HY134928AS5MG
Pyridinium bisretinoid A2E-d4 TFA is a deuterium-labeled analog of the retinal bisretinoid A2E supplied as the trifluoroacetate salt for use as an isotopically labeled standard in biochemical and phototoxicity research. It models the photoactivated behavior of native A2E and is intended for quantitative analysis, tracer experiments, and studies of apoptosis and autophagy in retinal pigment epithelium.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • Supplied as a trifluoroacetate salt for handling and solubility in common organic solvents.
  • Suitable for phototoxicity, apoptosis, and autophagy research applications.
  • Certificate of analysis available reporting purity by NMR (90.0%).
  • Offered in small research pack sizes appropriate for analytical workflows.