Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridinium Bromide Perbromide 85.0+%, TCI America™
SDP

Catalog No. P0825100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0825100G 100 g
P082525G 25 g
P0825500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0825100G Supplier TCI America Supplier No. P0825100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H6Br3N
MDL Number MFCD00013223
Synonym Pyridine Hydrobromide Perbromide, Pyridinium Tribromide

Specifications

Melting Point 136°C
Quantity 100 g
UN Number 1759
Formula Weight 319.82
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium Bromide Perbromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.