Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridinium Fluorochromate 98.0+%, TCI America™
SDP

Catalog No. P10885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10885G Supplier TCI America Supplier No. P10885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxidizing agent suitable for selective oxidation applications.

Chemical Identifiers

CAS 83042-08-4
Molecular Formula C5H6CrFNO3
Molecular Weight (g/mol) 199.1
MDL Number MFCD00151617
Synonym pyridin-1-ium; trioxochromium; hydrofluoride, chromium trioxide pyridium hydrofluoride, pyridin-1-ium; tris oxidanylidene chromium; hydrofluoride

Specifications

Quantity 5 g
UN Number 3087
Formula Weight 199.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium Fluorochromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.