Learn More
Aviva Systems Biology Corp Pyridinoline ELISA Kit (OKEH02582) | 96 Wells
SDP
Catalog No. NC1860411
Encompass
Aviva Systems Biology Pyridinoline ELISA Kit (OKEH02582) is based on a competitive enzyme immunoassay technique. The microtiter well-plate in this kit has been pre-coated with an anti-Pyridinoline antibody. Sample or standards are added to the wells along with a fixed quantity of biotinylated Pyridinoline and incubated. The Pyridinoline found in the sample or standards competes with the biotinylated Pyridinoline for limited binding sites on the immobilized anti-Pyridinoline antibody. Excess unbound biotinylated Pyridinoline and sample or standard Pyridinoline is washed from the plate. Avidin-HRP conjugate is added, incubated and washed. An enzymatic reaction is then produced through the addition of TMB substrate which is catalyzed by the immobilized HRP to generate a blue color product that changes yellow after adding acidic stop solution.
Catalog No. NC1860411 Supplier Aviva Systems Biology Corp Supplier No. OKEH02582
Aviva Systems Biology Pyridinoline ELISA Kit (OKEH02582) is based on a competitive enzyme immunoassay technique. The microtiter well-plate in this kit has been pre-coated with an anti-Pyridinoline antibody. Sample or standards are added to the wells along with a fixed quantity of biotinylated Pyridinoline and incubated. The Pyridinoline found in the sample or standards competes with the biotinylated Pyridinoline for limited binding sites on the immobilized anti-Pyridinoline antibody. Excess unbound biotinylated Pyridinoline and sample or standard Pyridinoline is washed from the plate. Avidin-HRP conjugate is added, incubated and washed. An enzymatic reaction is then produced through the addition of TMB substrate which is catalyzed by the immobilized HRP to generate a blue color product that changes yellow after adding acidic stop solution.