Learn More
Cayman Chemical PyrIdoal IsonIcotInyl hyd 50mg
SDP
Catalog No. 502984471
Encompass_Preferred
A lipophilic, nontoxic, iron-chelating agent that shows high iron chelation efficacy both in vitro in cell culture models and also in vivo in rats and mice
Catalog No. 50-298-4471 Supplier Cayman Chemical Supplier No. 1935750 MG
A lipophilic, nontoxic, iron-chelating agent that shows high iron chelation efficacy both in vitro in cell culture models and also in vivo in rats and mice