Learn More
Cayman Chemical PyrIdomycIn 2.5mg
SDP
Catalog No. 502980435
Encompass_Preferred
A structurally unusual antibiotic that inhibits NADH-dependent enoyl (Acyl-Carrier-Protein) reductase InhA, preventing mycolic acid synthesis in M. tuberculosis, including isoniazid-resistant strains (MICs = 0.31-0.63 UG/ml)
Catalog No. 50-298-0435 Supplier Cayman Chemical Supplier No. 170952.5 MG
A structurally unusual antibiotic that inhibits NADH-dependent enoyl (Acyl-Carrier-Protein) reductase InhA, preventing mycolic acid synthesis in M. tuberculosis, including isoniazid-resistant strains (MICs = 0.31-0.63 UG/ml)