Learn More
Medchemexpress LLC HY-14435 2mg , Pyridone 6 CAS:457081-03-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY144352MG
Encompass_Preferred
HY-14435 2mg Pyridone 6 CAS: 457081-03-7 Pyridone 6 is a pan-JAK inhibitor, which potently inhibits the JAK kinase family, with IC50s of 1 nM for JAK2 and TYK2, 5 nM for JAK3, and 15 nM for JAK1, while displaying significantly weaker affinities (130 nM to >10 mM) for other protein tyrosine kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0856