Learn More
Medchemexpress LLC Pyridostatin hydrochloride | 1781882-65-2 | MFCD28411715 | 100.0% | 778.94 g/mol | C31H37Cl5N8O5 | 10 MG
SDP
Catalog No. 5000286504
Encompass_Preferred
Pyridostatin hydrochloride is a small-molecule G-quadruplex (G4) stabilizing agent used in molecular biology and cancer research to induce replication- and transcription-dependent DNA damage and to probe G4 structures in DNA and RNA. It is supplied as a high-purity white to off-white solid and is used in vitro and in cell-based assays.
  • Stabilizes G-quadruplex structures in DNA and RNA.
  • Induces replication- and transcription-dependent DNA damage in cell models.
  • Enables study of genomic instability and G4-related biological processes.
  • Supplied as a high-purity solid suitable for analytical and biological assays.
  • Available in small-scale quantities and solution formulations for convenience.
  • Store solid at 4°C under inert atmosphere; store solutions at low temperatures.
Catalog No. 50-002-86504 Supplier Medchemexpress LLC Supplier No. HY15176A10MG
Pyridostatin hydrochloride is a small-molecule G-quadruplex (G4) stabilizing agent used in molecular biology and cancer research to induce replication- and transcription-dependent DNA damage and to probe G4 structures in DNA and RNA. It is supplied as a high-purity white to off-white solid and is used in vitro and in cell-based assays.
  • Stabilizes G-quadruplex structures in DNA and RNA.
  • Induces replication- and transcription-dependent DNA damage in cell models.
  • Enables study of genomic instability and G4-related biological processes.
  • Supplied as a high-purity solid suitable for analytical and biological assays.
  • Available in small-scale quantities and solution formulations for convenience.
  • Store solid at 4°C under inert atmosphere; store solutions at low temperatures.