Learn More
Cayman Chemical PyrIdostIgmIn-d3bromIde 10mg
SDP
Catalog No. 502910676
Encompass_Preferred
An internal standard for the quantification of pyridostigmine by GC- or LC-MS
Catalog No. 50-291-0676 Supplier Cayman Chemical Supplier No. 3520810 MG
An internal standard for the quantification of pyridostigmine by GC- or LC-MS