Learn More
Cayman Chemical Pyridostigmine (bromide), Purity: ≥98%, CAS: 101-26-8, 100 mg
SDP
Catalog No. 502995271
Encompass_Preferred
An inhibitor of AChE and BChE (IC50s = 0.35 and 1 μM, respectively, for the human enzymes); reduces organophosphate pesticide-induced decreases in AChE activity at 10 µM; reduces soman-induced membrane depolarization and decreases in AChE activity in isolated human muscle bundles in a concentration-dependent manner; prevents tachycardia and increases in sympathetic tone in mice following myocardial infarction induced by left coronary artery ligation at 3 mg/kg per day for four weeks
Catalog No. 50-299-5271 Supplier Cayman Chemical Supplier No. 23831100 MG
An inhibitor of AChE and BChE (IC50s = 0.35 and 1 μM, respectively, for the human enzymes); reduces organophosphate pesticide-induced decreases in AChE activity at 10 µM; reduces soman-induced membrane depolarization and decreases in AChE activity in isolated human muscle bundles in a concentration-dependent manner; prevents tachycardia and increases in sympathetic tone in mice following myocardial infarction induced by left coronary artery ligation at 3 mg/kg per day for four weeks