Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyridostigmine-d6 bromide | 2375858-08-3 | 100.0% | 267.15 | C9H7D6BrN2O2 | 10 MG
SDP

Catalog No. 5000298989
Encompass_Preferred

Pyridostigmine-d6 (bromide) is the deuterium-labeled form of pyridostigmine used as a stable-isotope research standard and a reversible cholinesterase inhibitor for biochemical and neuronal signaling studies.

  • Deuterium-labeled internal standard suitable for mass spectrometry and quantitative assays.
  • High chemical purity (99.96%).
  • Molecular weight 267.15; molecular formula C9H7D6BrN2O2.
  • CAS number 2375858-08-3 for unambiguous identification.
  • Available in small research pack sizes, including a 10 mg vial.
  • Storage: store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-98989 Supplier Medchemexpress LLC Supplier No. HYB0207AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyridostigmine-d6 (bromide) is the deuterium-labeled form of pyridostigmine used as a stable-isotope research standard and a reversible cholinesterase inhibitor for biochemical and neuronal signaling studies.

  • Deuterium-labeled internal standard suitable for mass spectrometry and quantitative assays.
  • High chemical purity (99.96%).
  • Molecular weight 267.15; molecular formula C9H7D6BrN2O2.
  • CAS number 2375858-08-3 for unambiguous identification.
  • Available in small research pack sizes, including a 10 mg vial.
  • Storage: store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.