Learn More
Medchemexpress LLC Pyridostigmine-d6 bromide | 2375858-08-3 | 100.0% | 5 MG
SDP
Catalog No. 5000407109
Encompass_Preferred
Pyridostigmine-d6 bromide is a deuterium-labeled Pyridostigmine, functioning as a parasympathomimetic and a reversible cholinesterase inhibitor. It is used as a tracer and an internal standard in quantitative analyses.
  • Deuterium-labeled compound
  • Functions as a parasympathomimetic and reversible cholinesterase inhibitor
  • Used as a tracer for quantitative analysis
  • Serves as an internal standard in analytical techniques like NMR, GC-MS, and LC-MS
  • Incorporates stable heavy isotopes
Catalog No. 50-004-07109 Supplier Medchemexpress LLC Supplier No. HYB0207AS5MG
Pyridostigmine-d6 bromide is a deuterium-labeled Pyridostigmine, functioning as a parasympathomimetic and a reversible cholinesterase inhibitor. It is used as a tracer and an internal standard in quantitative analyses.
  • Deuterium-labeled compound
  • Functions as a parasympathomimetic and reversible cholinesterase inhibitor
  • Used as a tracer for quantitative analysis
  • Serves as an internal standard in analytical techniques like NMR, GC-MS, and LC-MS
  • Incorporates stable heavy isotopes