Learn More
Apexbio Technology LLC Pyridoxal 5 phosphate 54-47-7 500mg
SDP
Catalog No. 5000088048
Encompass_Preferred
Pyridoxal 5-phosphate (CAS 54-47-7) is a biologically active derivative of vitamin B6 functioning primarily as a coenzyme in numerous enzymatic reactions It is designed to facilitate amino acid metabolism and biosynthesis pathways including neurotransmitter and sphingolipid synthesis as well as porphyrin biosynthesis Pyridoxal 5-phosphate exerts its biological activity primarily through coenzyme-mediated facilitation of transamination and other enzymatic transformations Its activity and affinity vary depending on the specific enzyme involved with reported enzyme IC50 values ranging broadly depending on enzyme type and experimental conditions Based on these pharmacological properties Pyridoxal 5-phosphate holds research potential in elucidating enzyme mechanisms metabolic pathway profiling assay optimization and the study of biosynthetic processes
Catalog No. 50-000-88048 Supplier Apexbio Technology LLC Supplier No. B7910500
Pyridoxal 5-phosphate (CAS 54-47-7) is a biologically active derivative of vitamin B6 functioning primarily as a coenzyme in numerous enzymatic reactions It is designed to facilitate amino acid metabolism and biosynthesis pathways including neurotransmitter and sphingolipid synthesis as well as porphyrin biosynthesis Pyridoxal 5-phosphate exerts its biological activity primarily through coenzyme-mediated facilitation of transamination and other enzymatic transformations Its activity and affinity vary depending on the specific enzyme involved with reported enzyme IC50 values ranging broadly depending on enzyme type and experimental conditions Based on these pharmacological properties Pyridoxal 5-phosphate holds research potential in elucidating enzyme mechanisms metabolic pathway profiling assay optimization and the study of biosynthetic processes