Learn More
Selleck Chemical LLC Pyridoxal 5-phosphate hydrate
SDP
Catalog No. 5000004220
Encompass_Preferred
Pyridoxal 5-phosphate (pyridoxal phosphate Vitamin B6 phosphate PAL-P PLP) a vitamin B6 phosphate is a coenzyme in a variety of enzymatic reactions Pyridoxal 5-phosphate at concentrations 0 5 mM inhibits of polymerization deoxynucleoside triphosphate catalysed by variety of DNA polymerases isolated from type C RNA tumor viruses The combination of Pyridoxal 5-phosphate and NEIL2 will make NEIL2 completely lose its ability to bind damaged DNA
Catalog No. 50-000-04220 Supplier Selleck Chemical LLC Supplier No. S61151G
Pyridoxal 5-phosphate (pyridoxal phosphate Vitamin B6 phosphate PAL-P PLP) a vitamin B6 phosphate is a coenzyme in a variety of enzymatic reactions Pyridoxal 5-phosphate at concentrations 0 5 mM inhibits of polymerization deoxynucleoside triphosphate catalysed by variety of DNA polymerases isolated from type C RNA tumor viruses The combination of Pyridoxal 5-phosphate and NEIL2 will make NEIL2 completely lose its ability to bind damaged DNA