Learn More
Medchemexpress LLC Pyridoxal 5'-phosphate hydrate | 853645-22-4 | 99.8% | 247.14 | 1 MG
SDP
Catalog No. 5000452432
Encompass_Preferred
Pyridoxal 5'-phosphate hydrate is the active form of vitamin B6 and an essential cofactor for various enzymes, including aromatic l-amino acid decarboxylase, which produces the neurotransmitters dopamine and serotonin. It is the most significant coenzyme variant in the intracellular phosphorylation of vitamin B6 and can be interconverted with other variants like pyridoxine 5′‐phosphate (PNP) and pyridoxamine 5′-phosphate (PMP).
  • Activates the ERK/c-Jun signaling pathway and promotes IGFBP1 protein expression.
  • Inhibits the proliferation of cancer cells.
  • Alleviates Aβ25-35-induced spatial and long-term memory impairment in mouse models.
  • Inhibits the formation of advanced glycation end-products (AGEs).
  • Prevents the progression of diabetic nephropathy.
Catalog No. 50-004-52432 Supplier Medchemexpress LLC Supplier No. HYW011727S1MG
Pyridoxal 5'-phosphate hydrate is the active form of vitamin B6 and an essential cofactor for various enzymes, including aromatic l-amino acid decarboxylase, which produces the neurotransmitters dopamine and serotonin. It is the most significant coenzyme variant in the intracellular phosphorylation of vitamin B6 and can be interconverted with other variants like pyridoxine 5′‐phosphate (PNP) and pyridoxamine 5′-phosphate (PMP).
  • Activates the ERK/c-Jun signaling pathway and promotes IGFBP1 protein expression.
  • Inhibits the proliferation of cancer cells.
  • Alleviates Aβ25-35-induced spatial and long-term memory impairment in mouse models.
  • Inhibits the formation of advanced glycation end-products (AGEs).
  • Prevents the progression of diabetic nephropathy.