Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyridoxal 5'-phosphate (hydrate) | 853645-22-4 | 99.8% | 247.14 | 10 G
SDP

Catalog No. 5000452428
Encompass_Preferred

Pyridoxal phosphate, also known as Pyridoxal 5′-phosphate, is the active form of vitamin B6 and an endogenous metabolite. It acts as an inhibitor of reverse transcriptases and is used for the treatment of tardive dyskinesia. Its biological activity includes inhibition of DNA Polymerase, RNase H, and NEIL2, as well as the proliferation of SK-OV-3 and OVCAR-3 cancer cells. In vivo, it alleviates memory impairment in mouse models and inhibits the formation of advanced glycation end-products (AGEs) in rat diabetes models.

  • Appearance: Off-white to light yellow solid
  • Purity (HPLC): 99.76%
  • Storage (solid): 4°C, sealed, away from moisture
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-52428 Supplier Medchemexpress LLC Supplier No. HYW01172710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyridoxal phosphate, also known as Pyridoxal 5′-phosphate, is the active form of vitamin B6 and an endogenous metabolite. It acts as an inhibitor of reverse transcriptases and is used for the treatment of tardive dyskinesia. Its biological activity includes inhibition of DNA Polymerase, RNase H, and NEIL2, as well as the proliferation of SK-OV-3 and OVCAR-3 cancer cells. In vivo, it alleviates memory impairment in mouse models and inhibits the formation of advanced glycation end-products (AGEs) in rat diabetes models.

  • Appearance: Off-white to light yellow solid
  • Purity (HPLC): 99.76%
  • Storage (solid): 4°C, sealed, away from moisture
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.