Learn More
Abcam Pyrido x al- 5'-phosphate monohydrate, Vitamin B6 metabolite, 25G
Supplier:  Abcam AB14622825G
Encompass_Preferred
MW 265.16 g/mol, Purity >98%. Vitamin B6 metabolite that acts as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase. Also serves as a coenzyme in some decarboxylation and deamination reactions. Can modify lysyl and valyl residues in proteins. Has the ability to inhibit purinergic receptors and intracellular influx of Ca²+.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4820