Learn More
Alkali Scientific Pyridoxal 5 Phosphate Monohydrate, 25 Grams
SDP
Catalog No. 5000661896
Encompass_Preferred
Pyridoxal 5-Phosphate Monohydrate is the active form of vitamin B6, used as an essential cofactor in numerous enzymatic reactions, including those involved in amino acid metabolism, neurotransmitter synthesis, and glycogen metabolism. Available in a 25-gram quantity, it is used in research on enzyme kinetics, metabolic disorders, and neurological diseases. Pyridoxal phosphate is vital for the proper function of enzymes that convert amino acids into other biochemicals essential for energy production and brain function. It is widely used in laboratory research on vitamin B6 deficiency, neurobiology, and metabolic pathways involving amino acids.
Catalog No. 50-006-61896 Supplier Alkali Scientific Supplier No. RID001CP
Pyridoxal 5-Phosphate Monohydrate is the active form of vitamin B6, used as an essential cofactor in numerous enzymatic reactions, including those involved in amino acid metabolism, neurotransmitter synthesis, and glycogen metabolism. Available in a 25-gram quantity, it is used in research on enzyme kinetics, metabolic disorders, and neurological diseases. Pyridoxal phosphate is vital for the proper function of enzymes that convert amino acids into other biochemicals essential for energy production and brain function. It is widely used in laboratory research on vitamin B6 deficiency, neurobiology, and metabolic pathways involving amino acids.