Learn More
Medchemexpress LLC Pyridoxal 5'-phosphate monohydrate | 41468-25-1 | 98.0% | 265.16 | 1 ML
SDP
Catalog No. 5000453681
Encompass_Preferred
Pyridoxal 5'-phosphate monohydrate, the active form of vitamin B6, is an essential cofactor for multiple enzymes. It plays a crucial role in the production of neurotransmitters like dopamine and serotonin and is interconvertible with other vitamin B6 variants.
  • Active form of vitamin B6
  • Essential cofactor for various enzymes
  • Involved in neurotransmitter production (dopamine and serotonin)
  • Activates the ERK/c-Jun signaling pathway in HepG2 cells
  • Inhibits proliferation of cancer cells SK-OV-3 and OVCAR-3
  • Alleviates Aβ25-35-induced memory impairment in mouse models
  • Inhibits advanced glycation end-products (AGEs) formation and prevents diabetic nephropathy progression in rat models
Catalog No. 50-004-53681 Supplier Medchemexpress LLC Supplier No. HYW011727A10MM/1ML
Pyridoxal 5'-phosphate monohydrate, the active form of vitamin B6, is an essential cofactor for multiple enzymes. It plays a crucial role in the production of neurotransmitters like dopamine and serotonin and is interconvertible with other vitamin B6 variants.
  • Active form of vitamin B6
  • Essential cofactor for various enzymes
  • Involved in neurotransmitter production (dopamine and serotonin)
  • Activates the ERK/c-Jun signaling pathway in HepG2 cells
  • Inhibits proliferation of cancer cells SK-OV-3 and OVCAR-3
  • Alleviates Aβ25-35-induced memory impairment in mouse models
  • Inhibits advanced glycation end-products (AGEs) formation and prevents diabetic nephropathy progression in rat models