Learn More
Alkali Scientific Pyridoxal 5 Phosphate Monohydrate, 5 Grams
SDP
Catalog No. 5000661910
Encompass_Preferred
Pyridoxal 5 Phosphate Monohydrate is a derivative of vitamin B6 that serves as an essential cofactor in numerous enzyme reactions within biochemical pathways. The 5-gram quantity of this reagent is ideal for small-scale research in enzyme catalysis and metabolic studies. Pyridoxal 5 phosphate is involved in amino acid metabolism, neurotransmitter synthesis, and the production of hemoglobin. It is also critical in studies investigating enzymatic reactions in pharmaceutical research and clinical diagnostics. As a high-purity reagent, it is commonly used in laboratories for research on vitamin B6-dependent enzymes, offering reliable results in studies related to human health and disease treatment.
Catalog No. 50-006-61910 Supplier Alkali Scientific Supplier No. RID002CP
Pyridoxal 5 Phosphate Monohydrate is a derivative of vitamin B6 that serves as an essential cofactor in numerous enzyme reactions within biochemical pathways. The 5-gram quantity of this reagent is ideal for small-scale research in enzyme catalysis and metabolic studies. Pyridoxal 5 phosphate is involved in amino acid metabolism, neurotransmitter synthesis, and the production of hemoglobin. It is also critical in studies investigating enzymatic reactions in pharmaceutical research and clinical diagnostics. As a high-purity reagent, it is commonly used in laboratories for research on vitamin B6-dependent enzymes, offering reliable results in studies related to human health and disease treatment.