Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxal-5-phosphate, Monohydrate, 97%, Spectrum™ Chemical
SDP

Catalog No. 18605988 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-988 1 kg Amber Glass Bottle
18-605-983 1 g Amber Glass Bottle
18-605-984 5 g Amber Glass Bottle
18-605-985 25 g Amber Glass Bottle
18-605-986 100 g Amber Glass Bottle
18-605-987 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-988 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PY1011KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

PY101, 41468-25-1, C8H10NO6P·H2O

Spectrum™ Chemical Pyridoxal-5-phosphate, Monohydrate

Chemical Identifiers

CAS 41468-25-1
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
IUPAC Name [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 97%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.