Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxal-5-Phosphate Monohydrate, MP Biomedicals™

Catalog No. MP210276925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210276925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210276925 Supplier MP Biomedicals, Inc Supplier No. 0210276925
Add to Cart
Add to Cart

Acts as a coenzyme in transamination, decarboxylation and deamination reactions. Pyridoxal-5-Phosphate Monohydrate, MP Biomedicals is an active form of Vitamin B6, an enzyme prosthetic group. It is useful for modifying lysyl and valyl residues in proteins

PLP acts as a coenzyme in conversion reactions, such as the conversion of dopa into dopamine, of glutamate into GABA, and of histidine to histamine. It works mechanistically through formation of a Schiff-base.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41468-25-1
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate
PubChem CID 38882
IUPAC Name [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

Melting Point 140°C to 143°C (literature)
Quantity 25 g
Solubility Information Soluble in 1M Hydrochloric acid (50mg/mL; clear and colorless to light yellow-green solution).
Formula Weight 265.1
Percent Purity ∼98%
Physical Form Powder
Recommended Storage Store at –20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.