Learn More
Cayman Chemical Pyridoxal 5'-phosphate (hydrate), Purity: ≥95%, CAS: 853645-22-4, 25 g
SDP
Catalog No. 502984687
Encompass_Preferred
The active form of vitamin B6, which serves as a cofactor for many different enzymes; used to study the pyridoxal 5′-phosphate-dependent active sites of these enzymes
Catalog No. 50-298-4687 Supplier Cayman Chemical Supplier No. 2035225 G
The active form of vitamin B6, which serves as a cofactor for many different enzymes; used to study the pyridoxal 5′-phosphate-dependent active sites of these enzymes