Learn More
Cayman Chemical PyrIdoxal 5-phosphatehydra 5g
SDP
Catalog No. 502984531
Encompass_Preferred
The active form of vitamin B6, which serves as a cofactor for many different enzymes; used to study the pyridoxal 5′-phosphate-dependent active sites of these enzymes
Catalog No. 50-298-4531 Supplier Cayman Chemical Supplier No. 203525 G
The active form of vitamin B6, which serves as a cofactor for many different enzymes; used to study the pyridoxal 5′-phosphate-dependent active sites of these enzymes