Learn More
Medchemexpress LLC Pyridoxal | 66-72-8 | MFCD00464806 | 100.0% | 167.16 | C8H9NO3 | 25 MG
SDP
Catalog No. 5000239755
Encompass_Preferred
Pyridoxal is an analytical standard of pyridoxal (a form of vitamin B6) provided as a high-purity reference material for research and analytical applications. It is intended for method development, calibration, and quantitative or qualitative assays.
  • High purity (99.97%) suitable for analytical use.
  • Supplied in small, well-defined packs; 25 MG available.
  • Compatible with HPLC, GC, and MS methods.
  • Molecular weight 167.16 and chemical formula C8H9NO3.
  • Useful for studies of vitamin B6 metabolism and related biochemical assays.
Catalog No. 50-002-39755 Supplier Medchemexpress LLC Supplier No. HY107469R25MG
Pyridoxal is an analytical standard of pyridoxal (a form of vitamin B6) provided as a high-purity reference material for research and analytical applications. It is intended for method development, calibration, and quantitative or qualitative assays.
  • High purity (99.97%) suitable for analytical use.
  • Supplied in small, well-defined packs; 25 MG available.
  • Compatible with HPLC, GC, and MS methods.
  • Molecular weight 167.16 and chemical formula C8H9NO3.
  • Useful for studies of vitamin B6 metabolism and related biochemical assays.