Learn More
Medchemexpress LLC Pyridoxal | 66-72-8 | MFCD00464806 | 100.0% | 167.16 g·mol^-1 | C8H9NO3 | 50 MG
SDP
Catalog No. 5000239756
Encompass_Preferred
Pyridoxal is the aldehyde form of vitamin B6 supplied as a high-purity analytical/research standard (CAS 66-72-8). It is used as a reference compound in biochemical and analytical applications and for studies of vitamin B6 metabolism. Molecular formula C8H9NO3; molecular weight 167.16 g·mol^-1. Supplied as a solid powder in small laboratory pack sizes.
  • High purity suitable for analytical and reference use.
  • Molecular formula C8H9NO3 and molecular weight 167.16 g·mol^-1.
  • Provided as a solid powder for easy handling and storage.
  • Useful in studies of vitamin B6 metabolism and enzymatic assays.
  • Available in small quantities for research-scale experiments.
Catalog No. 50-002-39756 Supplier Medchemexpress LLC Supplier No. HY10746950MG
Pyridoxal is the aldehyde form of vitamin B6 supplied as a high-purity analytical/research standard (CAS 66-72-8). It is used as a reference compound in biochemical and analytical applications and for studies of vitamin B6 metabolism. Molecular formula C8H9NO3; molecular weight 167.16 g·mol^-1. Supplied as a solid powder in small laboratory pack sizes.
  • High purity suitable for analytical and reference use.
  • Molecular formula C8H9NO3 and molecular weight 167.16 g·mol^-1.
  • Provided as a solid powder for easy handling and storage.
  • Useful in studies of vitamin B6 metabolism and enzymatic assays.
  • Available in small quantities for research-scale experiments.