Learn More
Medchemexpress LLC Pyridoxal | 66-72-8 | MFCD00464806 | 100.0% | 167.16 g/mol | C8H9NO3 | 50 MG
SDP
Catalog No. 5000241442
Encompass_Preferred
Pyridoxal is an analytical reference standard of vitamin B6 used in research and analytical laboratories for method development, calibration, and quality control in chromatographic and mass spectrometric assays. It is supplied as a small, high-purity solid suitable for quantitative and qualitative analyses.
Catalog No. 50-002-41442 Supplier Medchemexpress LLC Supplier No. HY107469R50MG
Pyridoxal is an analytical reference standard of vitamin B6 used in research and analytical laboratories for method development, calibration, and quality control in chromatographic and mass spectrometric assays. It is supplied as a small, high-purity solid suitable for quantitative and qualitative analyses.