Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyridoxal | 66-72-8 | MFCD00464806 | 99.6% | 167.16 g/mol | C8H9NO3 | 1 ML
SDP

Catalog No. 5000241052
Encompass_Preferred

Pyridoxal is a form of vitamin B6 used as a research reagent and endogenous metabolite. It is described as a neuroprotectant and is used in biochemical and metabolic studies. The material is supplied as a powder and as a ready-to-use 10 mM solution in DMSO (1 mL). For research use only.

  • High purity: 99.6%.
  • Molecular weight: 167.16 g/mol.
  • Available as powder or 10 mM solution in DMSO, 1 mL.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: biochemical assays and metabolic research.

Catalog No. 50-002-41052 Supplier Medchemexpress LLC Supplier No. HY10746910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyridoxal is a form of vitamin B6 used as a research reagent and endogenous metabolite. It is described as a neuroprotectant and is used in biochemical and metabolic studies. The material is supplied as a powder and as a ready-to-use 10 mM solution in DMSO (1 mL). For research use only.

  • High purity: 99.6%.
  • Molecular weight: 167.16 g/mol.
  • Available as powder or 10 mM solution in DMSO, 1 mL.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: biochemical assays and metabolic research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.