Learn More
Medchemexpress LLC Pyridoxal-d3 | 1173148-04-3 | MFCD11656098 | 98.7% | 170.18 | C8H6D3NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY107469S11MG
Encompass_Preferred
Pyridoxal-d3 is a deuterium-labeled form of pyridoxal (vitamin B6) used as a stable isotope tracer and internal standard in analytical and biochemical assays. The compound is a solid with molecular formula C8H6D3NO3, molecular weight 170.18, and reported purity of 98.71%, supplied in small quantities for reference and isotopic labeling applications.
  • Deuterium-labeled analogue for use as an internal standard in mass spectrometry and isotope tracing.
  • High purity suitable for quantitative analytical methods.
  • Small quantity packaging for reference material and method development.
  • Stable solid form for straightforward storage and handling.
Catalog No. 50-002-41042