Learn More
AdipoGen Pyridoxal hydrochloride
SDP
Supplier:  AdipoGen CDXP0118K001
Encompass_Preferred
Chemical. CAS 65-22-5. Formula C8H9NO3 . HCl. MW 203.62. Synthetic. For the labeling of amino acids and their detection in picomolar amounts.
Catalog No. 50-256-0247