Learn More
Research Products International Corp RPI Pyridoxal Hydrochloride, 500 Grams

Supplier: Research Products International Corp P14600500.0
- CAS Number: 65-22-5
- Molecular Formula: C8H9NO3 HCl
- Molecular Weight: 203.62 g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.