Learn More
Alkali Scientific Pyridoxal Hydrochloride, 5 Grams
SDP
Catalog No. 5000661566
Encompass_Preferred
Pyridoxal Hydrochloride is a vitamin B6 derivative widely used as a cofactor in enzymatic reactions involving amino acid metabolism. It plays an essential role in transamination, decarboxylation, and other biochemical processes that are critical for protein and neurotransmitter synthesis. This compound is commonly used in enzyme assays and metabolic studies to investigate vitamin-dependent reactions. Its hydrochloride salt form enhances stability and solubility in aqueous solutions, making it suitable for laboratory applications. Pyridoxal hydrochloride is frequently used in biochemical research, cell culture studies, and analytical procedures involving enzymatic activity and metabolic pathways.
Catalog No. 50-006-61566 Supplier Alkali Scientific Supplier No. YRI003CP
Pyridoxal Hydrochloride is a vitamin B6 derivative widely used as a cofactor in enzymatic reactions involving amino acid metabolism. It plays an essential role in transamination, decarboxylation, and other biochemical processes that are critical for protein and neurotransmitter synthesis. This compound is commonly used in enzyme assays and metabolic studies to investigate vitamin-dependent reactions. Its hydrochloride salt form enhances stability and solubility in aqueous solutions, making it suitable for laboratory applications. Pyridoxal hydrochloride is frequently used in biochemical research, cell culture studies, and analytical procedures involving enzymatic activity and metabolic pathways.