Learn More
Alkali Scientific Pyridoxal Hydrochloride, 500 Grams
SDP
Catalog No. 5000661567
Encompass_Preferred
Pyridoxal Hydrochloride is the hydrochloride form of Vitamin B6 and is utilized in various biochemical research applications, particularly in enzyme studies and metabolic research. It plays a critical role as a coenzyme in amino acid metabolism, neurotransmitter synthesis, and the formation of hemoglobin. This 500g package is ideal for laboratories requiring larger quantities of pyridoxal for ongoing research and large-scale applications. Pyridoxal Hydrochloride is also used in the production of pharmaceutical compounds and as a supplement in dietary research. It is essential in studies focused on the regulation of homocysteine levels and its effects on cardiovascular health.
Catalog No. 50-006-61567 Supplier Alkali Scientific Supplier No. YRI004CP
Pyridoxal Hydrochloride is the hydrochloride form of Vitamin B6 and is utilized in various biochemical research applications, particularly in enzyme studies and metabolic research. It plays a critical role as a coenzyme in amino acid metabolism, neurotransmitter synthesis, and the formation of hemoglobin. This 500g package is ideal for laboratories requiring larger quantities of pyridoxal for ongoing research and large-scale applications. Pyridoxal Hydrochloride is also used in the production of pharmaceutical compounds and as a supplement in dietary research. It is essential in studies focused on the regulation of homocysteine levels and its effects on cardiovascular health.