Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxal hydrochloride, MP Biomedicals™

Catalog No. ICN10276780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10276780 100 g
ICN10276701 1 g
ICN10276705 5 g
ICN10276725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10276780 Supplier MP Biomedicals, Inc Supplier No. 0210276780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 65-22-5 | C8H10ClNO3 | 203.622 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-22-5
Molecular Formula C8H10ClNO3
Molecular Weight (g/mol) 203.622
InChI Key FCHXJFJNDJXENQ-UHFFFAOYSA-N
Synonym pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
PubChem CID 6171
IUPAC Name 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
SMILES CC1=NC=C(C(=C1O)C=O)CO.Cl

Specifications

Color White
Quantity 100 g
Formula Weight 203.6
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxal hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.